Glossy White Polyester Barcode and Solar Panel Labels, 3" Core, Brady
Supplier: Brady Worldwide
Certificates
About this item
Permanent Polyester (B-423) is ideal for electronic PCB and component identification, bar code labels, rating plates, and solar panel identification.
- Aggressive adhesive
- Harsh environment, heat-resistant
- Permanent adhesive
Glossy white finish with permanent acrylic is thermal transfer printable. Service temperature of ‒70 to +110 °C (‒94 to +230 °F).
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, and THT 3" Core Printers.
Certifications: UL approved. CSA accepted. Meets requirements of a halogen-free material per DIN VDE 0472 part 815.
Ordering information: A ribbon is required to print this material. Use Part Number 89077-379 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 101076-574 for a different printer.