Nylon Cloth Wire and Lab Labels, 3" Core, Brady
Supplier: Brady Worldwide
Certificates
About this item
Nylon Cloth (B-499) wrap-around labels are highly pliable to conform to tightly curved surfaces such as wire and cable.
- Autoclavable material
- Suitable for cryogenic storage
- Service temperature of ‒40 to +90° C (‒40 to +193 °F)
Matte white polyamide coated nylon cloth material has a permanent acrylic adhesive and is thermal transfer printable. Recommended for indoor use.
Labels are designed for use with BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, or THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number 89077-374 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 94000-380 for a different printer.