High Adhesion Glossy Polyester with Acrylic Adhesive Labels, 3" Core, Brady
Supplier: Brady Worldwide
Certificates
About this item
Thermal transfer printable, glossy permanent polyester (B-422) labels are ideal for use in all commercial and industrial applications.
- Aggressive , heat-resistant, permanent adhesive
- Service temperature: ‒40 to +100° C (‒40 to +212° F)
Aggressive adhesive is ideal for powder-coated and textured flat surfaces, including low surface energy plastics. Designed to withstand exposure to numerous solvents.
Labels are designed for use with BBP®11 Label Printer, BBP®12 Label Printer, BBP®81 Label Printer, Brady IP® Series Label Printer, BradyPrinter i5100 Industrial Label Printer, BradyPrinter i7100 Industrial Label Printer, or THT 3" Core Printers.
Ordering information: A ribbon is required to print this material. Use Part Number 76251-721 for the BradyPrinter i5100 or BRADY IP® Series Label Printer; use 76460-980 for a different printer.