Axygen® Agarose, Low Melting Point, Corning
Supplier: Corning
Certificates
About this item
A high quality molecular biology grade agarose suitable for analytical and preparative electrophoresis of nucleic acids.
- Low melting temperatures and gel strength
- Electrophoresis of DNA fragments ≥1000 bp
- Suitable for tissue and cell culture and viral plaque assays
- Ideal for preparative electrophoresis
A low melting agarose that allows for the recovery of undamaged nucleic acids below their denaturation temperature.
In addition, in-gel manipulations can be performed without prior extraction of the DNA from the gel slice.
Specifications
- Catalog No:
- 75816-210
- Environmentally Preferable:
- Low Manufacturing Impact
- Gelling Temperature:
- ≥31 °C
- Melting Point:
- ≥76 °C
- Environmental Certification:
- I-RECs, ISO 14001 - Reynosa