- Pack type:Poly bottle
- Description:Differentiating solution
- Size:1 gal.
- Cat. No.:84000-110
- Application:Clear contrast between hematoxylin and eosin
- Supplier no.:74211
Epredia™ Differentiation Solution is a standard formulation of hydrochloric acid in denatured alcohol.
- Reproducible batch results allow for consistent staining in regressive staining procedures
- Solution provides clear differentiation and contrast between hematoxylin and eosin