ContactUsLinkComponent
NEBNext® Magnetic Separation Rack, New England Biolabs
NEBNext® Magnetic Separation Rack, New England Biolabs
Catalog # 76326-058
CAS Number:  
undefined
NEBNext® Magnetic Separation Rack, New England Biolabs
Catalog # 76326-058
Supplier Number:  S1515S
CAS Number:  
Specifications
  • Description:
    NEBNext® Magnetic separation rack
  • Size:
    24 tubes
  • Cat. No.:
    76326-058
Specifications
About this item
Next generation sequencing library preparation workflows include magnetic bead-based purification and size-selection steps. It is important for library yield and quality that bead separation be highly efficient and fast. This rack was designed for this application and contains rare earth Neodymium Iron Boron (NdFeB) magnets, the most powerful commercially available magnets, in an anodized aluminum rack. The rack holds 24×0.2 ml tubes, and is compatible with single tubes or strip tubes.
  • Fast separations in purification and size-selection steps in next generation sequencing workflows
  • Small-scale separation of magnetic particles
  • Anodized aluminum rack with Neodymium Iron Boron (NdFeB) rare earth magnets
  • 24 tube capacity: 8- and 12-strip 0.2 ml PCR tubes or individual 0.2 ml PCR tubes