ContactUsLinkComponent
Cytoblock™ Cell Block Preperation System, Epredia™
Cytoblock™ Cell Block Preperation System, Epredia™
Catalog # 102971-752
Supplier:  Epredia
CAS Number:  
undefined
Cytoblock™ Cell Block Preperation System, Epredia™
Catalog # 102971-752
Supplier:  Epredia
Supplier Number:  7401150
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Cytoblock™ cell block preparation system
  • Cat. No.:
    102971-752
  • Supplier no.:
    7401150
Specifications
About this item
Facilitate quick, reliable processing of FNAs, tissue fragments, and cell buttons with the Cytoblock™ Cell Block Preparation System.
  • Increases ability to process scanty or unseen material for cell block and reduces loss of critical material
  • Suitable for immunohistochemical techniques
  • Replaces time-consuming and costly agar and thrombin techniques