ContactUsLinkComponent
Centrifuge Tubes, RNase, DNase and Pyrogen Free, Globe Scientific
Centrifuge Tubes, RNase, DNase and Pyrogen Free, Globe Scientific
Catalog # 76478-224
Supplier:  Globe Scientific
undefined
Centrifuge Tubes, RNase, DNase and Pyrogen Free, Globe Scientific
Catalog # 76478-224
Supplier:  Globe Scientific
Supplier Number:  V-6285KG
Specifications
  • Cap material:
    HDPE
  • Cap type:
    Screw cap
  • Sterility:
    Sterile
  • Material:
    PP
  • Cat. No.:
    76478-224
  • Packaging:
    25/bag, 20 bags/case
  • Cap color:
    Blue
Specifications
About this item
These tubes are produced from polypropylene.
  • HDPE cap
  • Gamma sterile, RNase/DNase free
  • Max spin rate: 1000×g
They feature printed graduations and a white writing space.