ContactUsLinkComponent
DTM (Dermatophyte Test Medium) Agar, HardyFlask™, Hardy Diagnostics
DTM (Dermatophyte Test Medium) Agar, HardyFlask™, Hardy Diagnostics
Catalog # 76403-460
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
DTM (Dermatophyte Test Medium) Agar, HardyFlask™, Hardy Diagnostics
Catalog # 76403-460
Supplier:  Hardy Diagnostics
Supplier Number:  X15
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Pack type:
    Plastic flask
  • Description:
    Prepared media (DTM agar)
  • Size:
    20/pack
  • Cat. No.:
    76403-460
Specifications
About this item
Dermatophyte Test Medium (DTM) is a selective and differential medium recommended for the cultivation and isolation of pathogenic dermatophytic fungi.
  • Lightweight for reduced shipping cost and less chance of breakage
  • Stackable for increased conservation of space, both in incubator and refrigerator. No special storage racks are needed
  • Interlocking for increased stability when incubating and transporting
  • Large surface area for the inoculum
  • Slanted neck facilitates easy inoculation of the flat media surface
  • Improved clarity for better visibility of the growing culture, both front and back