ContactUsLinkComponent
Diamond Carelyte® Plus Electrolyte Analyzer, Diamond Diagnostics
  103881-356
undefined
Diamond Carelyte® Plus Electrolyte Analyzer, Diamond Diagnostics
  103881-356
 :  0-DD-CARELYTEPLUS
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Diamond Carelyte® Plus electrolyte analyzer
  • Cat. No.:
    103881-356
Electrolyte Analyzer, Carelyte® Plus, completely automated system measuring Na⁺, K⁺, Cl⁻, Ca⁺⁺ and in whole blood, serum, plasma and urine utilizing Ion Selective Electrode technology for veterinary purposes, designed with a powerful microprocessor allowing for easy interface with external LIS system.
  • 28 S analysis
  • 512 tests/hour
  • 40000 results stored
  • Touch screen display
  • Remote user access
  • Online service interface