Diamond SmartLyte® Plus Electrolyte Analyzer, Diamond Diagnostics
Diamond SmartLyte® Plus electrolyte analyzer
Catalog # 103881-354
Marketsource
Supplier: DIAMOND DIAGNOSTICS INC
Supplier #: 0-DD-SMARTLYTEPLUS
Diamond SmartLyte® Plus Electrolyte Analyzer, Diamond Diagnostics
Catalog # 103881-354
Supplier: DIAMOND DIAGNOSTICS INC
Marketsource
Jump to:
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
Diamond Smartlyte® Plus is an advanced electrolyte analyzer.
- 28 S Analysis
- 512 tests/hour
- 40000 results stored
- Touch screen display
- Remote user access
- Online service interface
The Diamond Diagnostics SmartLyte® Plus electrolyte analyzer is a completely automated system measuring Na⁺, K⁺, Cl⁻, Ca⁺ and Li⁺ in whole blood, serum, plasma and urine utilizing Ion Selective Electrode (ISE) technology. The Diamond SmartLyte® Plus is uniquely designed with a powerful microprocessor allowing for easy interface with external LIS systems, operation with a new touch screen interface, bar code scanner and storage and retrieval of over 10000 patient results. The system currently operates in over 12 languages and offers laboratories the option to utilize the analyzer for either human or veterinary applications.
Documents
Specifications
- Description:Diamond SmartLyte® Plus electrolyte analyzer
- Cat. No.:103881-354
Accessories
Product Information
- Catalog #For use withAvailabilityPrice
- Fluid packFluid pack103881-358Fluid Pack, ISE, for Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Startlyte V and Roche 9180,9181. Startlyte V and Roche 9180,9181, Compatible with: CareLyte, Alfa Wassermann Starlyte 5, Roche 9180 without chip.Log in for availabilityEach - $132.00
Product Information
- Catalog #For use withAvailabilityPrice
- Electrode conditioning solutionElectrode conditioning solution103881-360Electrode Conditioning Solution for Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Alfa Wassermann Starlyte III, Starlyte V, Idexx Vetlyte, Roche 9110, 9120, 9130, 9140, 9180, 9181.Log in for availabilityEach - $30.00
Product Information
- Catalog #For use withAvailabilityPrice
- K⁺ electrodeK⁺ electrode103881-374K⁺ Electrode for use on Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, , Idexx VetLyte, Alfa Wassermann Starlyte III, Starlyte V, Roche 9110, 9120, 9130, 9140, 9180, 9181.Log in for availabilityEach - $297.00
Product Information
- Catalog #For use withAvailabilityPrice
- Printer paperPrinter paper103881-386Printer paper for use on Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Idexx VetLyte, Alfa Wassermann Starlyte III, Starlyte V, Roche 9110, 9120, 9130, 9140, 9180, 9181.Log in for availabilityEach - $16.40
Product Information
- Catalog #For use withAvailabilityPrice
- Peristaltic pump tubingPeristaltic pump tubing103881-388Peristaltic Pump Tubing for use on Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus,Starlyte III + V, Idexx Vetlyte, Roche 9110, 9120, 9130, 9140, 9180, 9181 and moreLog in for availabilityEach - $59.00
Product Information
- Catalog #For use withAvailabilityPrice
- Sample sensorSample sensor103881-390Sample Sensor for Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Alfa Wassermann Starlyte V, Idexx Vetlyte, and Roche 9110, 9120, 9130, 9140, 9180, 9181.Log in for availabilityEach - $351.00
Product Information
- Catalog #For use withAvailabilityPrice
- Pinch valve tubing kitPinch valve tubing kit103881-392Pinch Valve Tubing Kit for SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Roche 9180, 9181, Idexx VetlyteLog in for availabilityEach - $231.00
Product Information
- Catalog #For use withAvailabilityPrice
- Sample probeSample probe103881-394Sample Probe for: Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Alfa Wassermann Starlyte III + V, Idexx Vetlyte and Roche 9120-9180.Log in for availabilityEach - $123.08
Product Information
- Catalog #For use withAvailabilityPrice
- ISE cleaning solutionISE cleaning solution103881-362ISE Cleaning Solution for Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Alfa Wassermann Starlyte III, Starlyte V, Idexx Vetlyte, Roche 9110, 9120, 9130, 9140, 9180, 9181,Log in for availabilityEach - $30.00
Product Information
- Catalog #For use withAvailabilityPrice
- Deproteinizer solutionDeproteinizer solution103881-364Deproteinizer Solution for Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Alfa Wassermann Starlyte III, Starlyte V, Idexx Vetlyte, Roche 9110, 9120, 9130, 9140, 9180, 9181Log in for availabilityEach - $30.00
Product Information
- Catalog #For use withAvailabilityPrice
- Urine diluentUrine diluent103881-366Urine Diluent for Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Alfa Wassermann Starlyte III, Starlyte V, Idexx Vetlyte, Roche 9110, 9120, 9130, 9140, 9180, 9181,Log in for availabilityEach - $30.00
Product Information
- Catalog #For use withAvailabilityPrice
- Na⁺ electrodeNa⁺ electrode103881-372Na⁺ Electrode for use on Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Alfa Wassermann Starlyte III, Starlyte V, Idexx Vetlyte and Roche 9110, 9120, 9130, 9140, 9180, 9181 .Log in for availabilityEach - $349.23
Product Information
- Catalog #For use withAvailabilityPrice
- Ca⁺⁺ electrodeCa⁺⁺ electrode103881-376Ca⁺⁺ Electrode for use on Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Alfa Wassermann Starlyte V, and Roche 9140, 9180, 9181.Log in for availabilityEach - $297.00
Product Information
- Catalog #For use withAvailabilityPrice
- Cl⁻ electrodeCl⁻ electrode103881-378Cl⁻ Electrode for Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Alfa Wassermann Starlyte III, Starlyte V, Idexx Vetlyte and Roche 9130, 9180, 9181.Log in for availabilityEach - $349.23
Product Information
- Catalog #For use withAvailabilityPrice
- Li⁺ electrodeLi⁺ electrode103881-380Li⁺ Electrode for Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Alfa Wassermann Starlyte V, and Roche 9180, 9181.Log in for availabilityEach - $297.00
Product Information
- Catalog #For use withAvailabilityPrice
- Reference electrodeReference electrode103881-382Compatible with: Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Roche 9110, 9120, 9130, 9140, 9180, 9181, Alfa Wassermann Starlyte III, Starlyte V, Idexx VetLyte, Diamond CareLyte, Alfa Wassermann ACE, ACEAlera, Axcel, NExCt, VetACELog in for availabilityEach - $358.20
Product Information
- Catalog #For use withAvailabilityPrice
- Reference electrode housingReference electrode housing103881-384Reference Electrode Housing for Diamond SmartLyte®, Smartlyte® Plus, Carelyte®, Carelyte® Plus, Alfa Wassermann Starlyte III, Starlyte V, Idexx Vetlyte, and Roche 9110, 9120, 9130, 9140, 9180, 9181.Log in for availabilityEach - $220.00