DNA (Deoxyribonucleic acid) (human genomic)
Catalog # PAG1521
Supplier: Promega Corporation
CAS Number:
Specifications
- Size:100 μg
- Cat. No.:PAG1521
- Supplier no.:G1521
Specifications
About this item
:
Storage Temperature:Refrigerator
CAS Number:9007-49-2
:73049-39-5
MDL Number:MFCD00130922