Order Entry
Puerto Rico
Orders LinkContactUsLinkComponent
CompactDry™ <i>E. coli</i> (EC)

CompactDry™ E. coli (EC)

CompactDry™ <i>E. coli</i> (EC)
Certificates
CompactDry™ <i>E. coli</i> (EC)

1 Options Available Below

  • Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
  • Return Policy
Product Details & Documents

CompactDry™ EC is a ready-to-use test method recommended for the isolation and enumeration of coliforms and Escherichia coli in raw materials, finished products, or on environmental surfaces pertaining to food and related industries.

  • Sturdy frame, stackable packaging
  • Self-diffusing media
  • Ability to subculture without damage to colonies
  • No leakage
  • Easy to label

CompactDry™ EC contains two kinds of chromogenic enzyme substrates: Magenta-Gal and X-Gluc. E. coli forms blue colonies, coliforms turn pink-purple. The total coliform group count is the sum of both the pink-purple and blue colonies.

Each cassette comes with four connected plates that can be easily snapped apart to support a variety of dilution scenarios.

Caution: Stored at room temperature vs the competitor products with storage at temperatures less than or equal to 8 °C.

Specifications for Product Family

Specifications

Specifications

  • Catalog No:
  • 10145-954
  • 76414-990
  • Storage Temperature:
  • 2 to 30 °C away from direct light
  • 2 to 30 °C away from direct light

Specifications

Product Family Options

Product Information

  • Includes
    Size
    Availability
    Price
  • 10145-954
    CompactDry™ E. coli (EC) and coliforms cassettes for colony counting
    CompactDry™ E. coli (EC) and coliforms cassettes for colony counting
    25 Packs
    240 plates/pack
    Discontinued
    Supplier Number: 54082
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    Product available on GSA Advantage®
    More
    Specifications
    Cat. No.
    10145-954
    Storage Temperature
    2 to 30 °C away from direct light
    Product available on GSA Advantage®
  • 76414-990
    CompactDry™ E. coli (EC) and coliforms cassettes for colony counting
    CompactDry™ E. coli (EC) and coliforms cassettes for colony counting
    25 Packs
    240 plates/pack
    Log in for availability
    Log in for availability
    Each
    Each - $304.00
    $304.00
    Each
       $304.00
    Minimum order quantity is 1

    Supplier Number: 54082
    Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
    Specifications:
    More
    Specifications
    Cat. No.
    76414-990
    Storage Temperature
    2 to 30 °C away from direct light
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.