Order Entry
Puerto Rico
ContactUsLinkComponent
Agar Deep, 1.5% (Water Agar), Hardy Diagnostics
Agar Deep, 1.5% (Water Agar), Hardy Diagnostics
Catalog # 76334-636
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
Agar Deep, 1.5% (Water Agar), Hardy Diagnostics
Catalog # 76334-636
Supplier:  Hardy Diagnostics
Supplier Number:  Q01
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Pack type:
    Tube
  • Description:
    Prepared media (agar deep, 1.5%)
  • Size:
    20/pack
  • Cat. No.:
    76334-636
  • Tube size:
    16×125 mm

Specifications

About this item

Hardy Diagnostics Agar, 1.5% (Water Agar) is used in the Kirby-Bauer alternative agar-overlay method of inoculation for antimicrobial disk diffusion susceptibility testing and for microbiological procedures that require an additive-free solid medium, such as in performing colony counts, testing antimicrobial disk potency, or in performing the modified fungal slide culture method.