ContactUsLinkComponent
KoolPlate™ Cooling Tray, Simport
KoolPlate™ Cooling Tray, Simport
Catalog # 76479-186
Supplier:  Simport Scientific
undefined
KoolPlate™ Cooling Tray, Simport
Catalog # 76479-186
Supplier:  Simport Scientific
Supplier Number:  M479-24
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
  • Description:
    KoolPlate™ 6 cooling trays with 1 insulator base
  • Dimensions:
    11×5.5×1"
  • Cat. No.:
    76479-186
Specifications
About this item
The Simport KoolPlate™ is ideal for chilling and preparing paraffin blocks, for sectioning at the microtome or for performing other laboratory procedures.
  • For chilling and preparing paraffin blocks
  • Keeps paraffin blocks cold up to 4 hours
  • Tray made of high impact polystyrene for longevity
  • Insulator base made of styrofoam
The KoolPlate™ freezes overnight (16 hours) in a conventional freezer and the surface remains cold for up to 4 hours when used with the supplied styrofoam insulator base placed under the KoolPlate™. It will prolong cooling and prevent moisture accumulation. It is made of thick wall high impact polystyrene in order to ensure a sturdy construction that will last for years.
The KoolPlate™ is stackable, a great space-saving feature. Warning–Not for use with organic solvents such as toluene, xylene, acetone, etc.
Caution: Not for use with organic solvents such as toluene, xylene, acetone, etc.