Order Entry
Puerto Rico
ContactUsLinkComponent
Sodium azide solution 10% (w/v), sterile
Toxic
Sodium azide solution 10% (w/v), sterile
Catalog # 101449-096
Supplier:  Teknova
CAS Number:  
Toxic
Sodium azide solution 10% (w/v), sterile
Catalog # 101449-096
Supplier:  Teknova
Supplier Number:  S0209
CAS Number:  

Specifications

  • Size:
    100 ml
  • Cat. No.:
    101449-096
  • Supplier no.:
    S0209

Specifications

About this item

 :
 :65.01 g/mol
UN:1687
ADR:6.1,II
 :GHS06
CAS Number:26628-22-8
Formula:NaN₃
MDL Number:MFCD00003536