ContactUsLinkComponent
ABS® Undercounter Pharmacy Freezers, Built-in Premier Series, Horizon Scientific
undefined
ABS® Undercounter Pharmacy Freezers, Built-in Premier Series, Horizon Scientific
  • Catalog No:
  • 76478-928
  • 76478-926
  • Model:
  • PH-ABT-HC-UCBI-0420A
  • PH-ABT-HC-UCBI-0420
  • Environmentally Preferable:
  • Safer Human and Environmental Health, Energy Efficient
  • Safer Human and Environmental Health, Energy Efficient
  • Cat. No.:
  • 76478-928
  • 76478-926
  • Refrigerant:
  • HFC-free refrigerant
  • HFC-free refrigerant
  • Display:
  • Digital
  • Digital
  • Defrost:
  • Auto
  • Manual
  • Electrical:
  • 115 V, 60 Hz, 1.8 A, 1/8 HP
  • 115 V, 60 Hz, 1.5 A, 1/8 HP
  • Shelves/drawers:
  • 3 shelves (adjustable)
  • 2 shelves (fixed)
  • Door Type:
  • 1 solid swing door, right hinged
  • 1 solid swing door, right hinged
  • Door Lock:
  • Keyed
  • Keyed
  • Access Ports:
  • ³/₈" probe
  • ³/₈" probe
  • Environmental Certification:
  • Energy Star certified
  • Weight:
  • 65 kg (143 lbs.)
  • 60 kg (132 lbs.)