TE Buffers 100X, G-Biosciences
Supplier: G-Biosciences
Certificates
About this item
TE Buffer (Tris-EDTA Buffer) is a common molecular biology buffer that is used to protect DNA and RNA from degradation. The Tris buffering agent and EDTA metal chelating properties help protect DNA and RNA.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.