Anti-S1 protein Hybridoma Polyclonal Antibody
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Type: Primary
Antigen: S1 protein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Hybridoma
Isotype: Mouse IgG1 kappa
Reactivity:
Specifications
- Catalog No:
- 103889-608
- 103889-606
- Antigen Symbol:
- S1 protein
- S1 protein
- Antigen Name:
- S1 protein
- S1 protein
- Antigen Synonyms:
- Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1
- Spike,S1 protein,Spike glycoprotein Subunit1,S glycoprotein Subunit1,Spike protein S1
- Antibody Type:
- Primary
- Primary
- Clonality:
- Polyclonal
- Polyclonal
- Conjugation:
- Unconjugated
- Unconjugated
- ELISA:
- Yes
- Yes
- Form:
- Powder
- Powder
- Purification:
- Affinity purified
- Affinity purified
- Cross Adsorption:
- This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. No cross-reactivity is detected with Spike protein RBD domain of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.
- This product is a specific antibody against SARS-CoV-2 Spike protein RBD domain. No cross-reactivity is detected with Spike protein RBD domain of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-NL63, HCoV-OC43 and HCoV-HKU1.
- Storage Temperature:
- For long term storage, the product should be stored at lyophilized state at -20° C or lower.
- For long term storage, the product should be stored at lyophilized state at -20° C or lower.
- Shipping Temperature:
- RT
- RT