Order Entry
Puerto Rico
ContactUsLinkComponent
SARS-CoV-2 Recombinant S protein RBD (from HEK293), Unconjugated
SARS-CoV-2 Recombinant S protein RBD (from HEK293), Unconjugated
Certificates
SARS-CoV-2 Recombinant S protein RBD (from HEK293), Unconjugated
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    SARS-CoV-2
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Anti-SARS-CoV-2 RBD Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind SARS-CoV-2 S protein RBD (F490L), His Tag (Cat. No. SPD-C52Hf) with an affinity constant of 53.8 nM as determined in a SPR assay (Biacore 8K) (Routinely tested).
    ; Loaded Human ACE2, Fc Tag (Cat. No. AC2-H5257) on Protein A Biosensor, can bind SARS-CoV-2 S protein RBD (F490L), His Tag (Cat. No. SPD-C52Hf) with an affinity constant of 57.9 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
    ; Loaded Anti-SARS-CoV-2 RBD Neutralizing Antibody, Human IgG1 (Cat. No. SAD-S35) on AHC Biosensor, can bind SARS-CoV-2 S protein RBD (F490L), His Tag (Cat. No. SPD-C52Hf) with an affinity constant of 45.2 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    QHD43416.1(F490L)
  • Protein Synonyms:
    Spike,S protein RBD,Spike glycoprotein Receptor-binding domain,S glycoprotein RBD,Spike protein RBD
  • Protein/Peptide Name:
    S protein RBD
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    26.6 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications