Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant BCMA (from HEK293), PE Labeled Protein
Human Recombinant BCMA (from HEK293), PE Labeled Protein
Certificates
Human Recombinant BCMA (from HEK293), PE Labeled Protein

Specifications

  • Species:
    Human
  • Storage Conditions:
    –20 °C, aviod light
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS,pH7.4,BSA.; 1e6 of the Anti-BCMA CAR-293 cells were stained with 100 μL of 1:50 dilution (2 μL stock solution in 100 μL FACS buffer) of PE-Labeled Human BCMA Protein, His Tag (Cat. No. BCA-HP2H2) and negative control protein respectively, PE signal was used to evaluate the binding activity (QC tested).
  • Gene ID:
    Q02223-1;608;NP_001183.2
  • Protein Synonyms:
    TNFRSF17,CD269,BCM,BCMA
  • Protein/Peptide Name:
    BCMA
  • Molecular Weight:
    10 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS,pH7.4,BSA.
  • Shipping Temperature:
    RT

Specifications