SARS-CoV-2 Recombinant S protein (from HEK293), Precision Avi
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Species:SARS-CoV-2
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS. Normally trehalose is added as protectant before lyophilization.
- Gene ID:QHD43416.1 (D614G)
- Protein Synonyms:Spike,S protein,Spike glycoprotein,S glycoprotein
- Protein/Peptide Name:S protein
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:139.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT