Human Recombinant NOTCH1 (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Human Jagged 1, His Tag (Cat. No. JA1-H52H9) immobilized on CM5 Chip can bind Human NOTCH1, Fc Tag (Cat. No. NO1-H5255) with an affinity constant of 5.13 μM as determined in a SPR assay (Biacore T200) (Routinely tested).; Loaded Human NOTCH1, Fc Tag (Cat. No. NO1-H5255) on Protein A Biosensor, can bind Human DLL4, His Tag (Cat. No. DL4-H5227) with an affinity constant of 67.5 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Human NOTCH1, Fc Tag (Cat. No. NO1-H5255) on Protein A Biosensor, can bind Human DLL1, His Tag (Cat. No. DL1-H52H8) with an affinity constant of 290 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:P46531-1;4851;NP_060087.3
- Protein Synonyms:NOTCH1,Notch 1,hN1,TAN1
- Protein/Peptide Name:NOTCH1
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:80.0 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT