Mouse Recombinant TGF-beta RII (from HEK293), Precision Avi
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Species:Mouse
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:Q62312-1
- Protein Synonyms:TGFBR2,TGFR2,TbetaR-II,TGFβR2
- Protein/Peptide Name:TGF-beta RII
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:21.7 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT