Human Recombinant TGF-beta 3 (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Biological Activity:Please contact us for detailed information.
- Gene ID:P10600-1
- Protein Synonyms:TGFB3,ARVD,TGF-beta3
- Protein/Peptide Name:TGF-beta 3
- Molecular Weight:14.4 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT