Human Recombinant IL-2 R beta & IL-2 R alpha & IL-2 R gamma (from HEK293), Unconjugated
Supplier: ACRO Biosystems
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Human IL-2RB&IL-2RA&IL-2RG, Fc Tag&Fc Tag (Cat. No. ILG-H5257) captured on CM5 chip via Anti-human IgG Fc antibodies surface can bind Human IL-2, Tag Free (Cat. No. IL2-H4113) with an affinity constant of 40.6 pM as determined in a SPR assay (Biacore 8K) (Routinely tested).
- Gene ID:P14784-1 (IL-2RB) & P01589-1 (IL-2RA) & P31785-1 (IL-2RG);3560 | 3559 | 3561;NP_000869.1 (IL-2RB) & NP_000408.1 (IL-2RA) & AAH14972.1 (IL-2RG)
- Protein/Peptide Name:IL-2 R beta & IL-2 R alpha & IL-2 R gamma
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:51.1 kDa | 75.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT
Specifications
Frequently Bought Together