Cynomolgus Recombinant MARCO (from HEK293), Unconjugated
- Conjugation:Unconjugated
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:A0A2K5X0Z2-1;102137861;XP_005573011.1
- Protein Synonyms:MARCO, SCARA2, Macrophage receptor with collagenous structure, Scavenger receptor class A member 2
- Protein/Peptide Name:MARCO
- Purity:>95% as determined by SDS-PAGE.
- Molecular Weight:46.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT