Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL-31 RA (from HEK293), Unconjugated
Human Recombinant IL-31 RA (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
Human Recombinant IL-31 RA (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    Q8NI17-1
  • Protein Synonyms:
    IL-31 RA,IL-31RA,hGLM-R,CRL3,GPL,ZcytoR17,GLM-R,IL-31R-alpha,Gp130-like receptor
  • Protein/Peptide Name:
    IL-31 RA
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    59.0 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together