Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IL-11 R alpha (from HEK293), Unconjugated
Human Recombinant IL-11 R alpha (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
Human Recombinant IL-11 R alpha (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Loaded Human IL-11, His Tag (Cat. No. IL1-H5243) on HIS1K Biosensor, can bind Human IL-11 R alpha, Fc Tag (Cat. No. ILR-H5256) with an affinity constant of 0.177 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
  • Gene ID:
    Q14626-1;3590;NP_001136256.1
  • Protein Synonyms:
    IL-11 R alpha,IL-11 RA,IL11RA,Interleukin-11 receptor subunit alpha,IL-11 receptor subunit alpha,IL-11R subunit alpha,IL-11R-alpha
  • Protein/Peptide Name:
    IL-11 R alpha
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    64.3 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together