Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant KRAS (from Insect Cells), Unconjugated, Acro Biosystems
Human Recombinant KRAS (from Insect Cells), Unconjugated, Acro Biosystems
Supplier:  ACRO Biosystems
Certificates
Human Recombinant KRAS (from Insect Cells), Unconjugated, Acro Biosystems
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 0.5 M NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P01116-2;3845;NP_004976.2
  • Protein Synonyms:
    GTPase Kras,K-Ras 2,Ki-Ras,c-K-ras,c-Ki-ras,KRAS2,RASK2,C-K-RAS,CFC2,K-RAS2A,K-RAS2B,K-RAS4A,K-RAS4B,KI-RAS,KRAS1,KRAS2,NS,NS3,RASK2,KRAS
  • Protein/Peptide Name:
    KRAS
  • Purity:
    >90% as determined by SDS-PAGE.
  • Molecular Weight:
    21.3 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in 50 mM Tris, 0.5 M NaCl, pH7.5. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together