Order Entry
Puerto Rico
ContactUsLinkComponent
Human Recombinant IFN-alpha 2b (from HEK293), Unconjugated
Human Recombinant IFN-alpha 2b (from HEK293), Unconjugated
Supplier:  ACRO Biosystems
Certificates
Human Recombinant IFN-alpha 2b (from HEK293), Unconjugated
Supplier:  ACRO Biosystems

Specifications

  • Conjugation:
    Unconjugated
  • Species:
    Human
  • Storage Conditions:
    –20 °C
  • Endotoxin Content:
    Less than 1.0 EU per μg by the LAL method.
  • Biological Activity:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Gene ID:
    P01563-1 (K46R);3440;NP_000596.2
  • Protein Synonyms:
    IFN-alpha 2b,IFNA2B,IFN-alpha 2 (alpha 2b),interferon alpha 2b
  • Protein/Peptide Name:
    IFN-alpha 2b
  • Purity:
    >95% as determined by SDS-PAGE.
  • Molecular Weight:
    45.7 kDa
  • Endotoxin Level:
    Low
  • Formulation:
    Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
  • Shipping Temperature:
    RT

Specifications

Frequently Bought Together