Human Recombinant IL-17A & IL-17F (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. Avantor will not lift restrictions for residential shipping addresses.
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.; Loaded Human IL-17 RA, Fc Tag (Cat. No. ILA-H5257) on Protein A Biosensor, can bind Human IL-17A&IL-17F Heterodimer Protein, Twin Strep&His Tag (Cat. No. ILF-H52W6) with an affinity constant of 3.70 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).; Loaded Human IL-17A&IL-17F Heterodimer Protein, Twin Strep&His Tag (Cat. No. ILF-H52W6) on NTA Biosensor, can bind Human IL-17 RC, Fc Tag (Cat. No. ILC-H5259) with an affinity constant of 92.3 nM as determined in BLI assay (ForteBio Octet Red96e) (Routinely tested).
- Gene ID:Q16552-1 (IL-17A) & Q96PD4-1 (IL-17F);3605 ∣ 112744;NP_002181.1 (IL-17A) & NP_443104.1 (IL-17F)
- Protein Synonyms:IL-17A,Interleukin-17A,CTLA-8,IL-17,IL-17F,Interleukin-17F,Cytokine ML-1
- Protein/Peptide Name:IL-17A & IL-17F
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:18.5 kDa ∣ 16.8 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT