SARS-CoV-2 Recombinant Nucleocapsid protein (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:SARS-CoV-2
- Storage Conditions:–20 °C
- Endotoxin Content:Less than 1.0 EU per μg by the LAL method.
- Biological Activity:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Gene ID:QHO62115.1 (R203K, G204R)
- Protein Synonyms:Nucleocapsid protein,NP,Protein N
- Protein/Peptide Name:Nucleocapsid protein
- Purity:>90% as determined by SDS-PAGE.
- Molecular Weight:47.4 kDa
- Endotoxin Level:Low
- Formulation:Lyophilized from 0.22 μm filtered solution in PBS, pH7.4. Normally trehalose is added as protectant before lyophilization.
- Shipping Temperature:RT