Human Recombinant IL4I1 (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Human
- Storage Conditions:–70℃
- Biological Activity:Please contact us for detailed information.
- Gene ID:Q96RQ9-1
- Protein Synonyms:L-amino-acid oxidase,Interleukin 4 induced protein 1, IL4I1
- Protein/Peptide Name:IL4I1
- Molecular Weight:62.6 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:Dry Ice