Cynomolgus Recombinant PVRIG (from HEK293), Unconjugated
Supplier: ACROBIOSYSTEMS INC MS
Certificates
About this item
Specifications
- Conjugation:Unconjugated
- Species:Cynomolgus monkey
- Storage Conditions:–20 °C
- Biological Activity:Please contact us for detailed information.
- Gene ID:A0A2K5WVV8-1
- Protein Synonyms:C7orf15,C7orf15MGC138295,CD112R,MGC104322,MGC138297,MGC2463,PVRIG,CD112 receptor
- Protein/Peptide Name:PVRIG
- Molecular Weight:40.5 kDa
- Endotoxin Level:Low
- Formulation:Please contact us for detailed information.
- Shipping Temperature:RT