Order Entry
Puerto Rico
ContactUsLinkComponent
 
L(+)-Histidine 98.5-101.5% (dried basis) USP, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™
 
L(+)-Histidine 98.5-101.5% (dried basis) USP, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., Macron Fine Chemicals™
 

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

 

  • USP - Assay (C H N 0 ) (dried basis):
    98.5 - 101.5%
  • USP - Identification:
    Passes Test
  • USP - Specific Rotation [A)25 AO (+):
    12.6 - 1.4.0°
  • USP - pH:
    7.0 - 8.5
  • USP - Loss on Orying at 105 °C:
    ≤0.2%
  • USP - Residue on Ignition:
    ≤0..4%
  • USP - Chloride (Cl):
    ≤0.05%
  • USP - Sulfate (S04):
    ≤0.03%
  • USP - Iron (Fe):
    ≤0.003%
  • USP - Related Compounds - Individual Impurities:
    ≤0.5%
  • USP - Related Compounds - Total Impurities:
    ≤2.0%
  • EP/BP - Assay (C H N 0 ) (dried basis):
    98.5 - 101.0%
  • EP/BP - Identification A:
    Passes Test
  • EP/BP - Identification B:
    Passes Test
  • EP/BP - Appearance of Solution:
    Passes Test
  • EP/BP - Specific Rotation [A)2°AO (+):
    11..4 - 12..4°
  • EP/BP-Ninhydrin-Positive Substances-Each:
    ≤0.2%
  • EP/BP-Ninhydrin-Positive Substances-Total Impurities:
    ≤0.5%
  • EP/BP - Chloride (Cl):
    ≤200 ppm
  • EP/BP - Sulfate (S04):
    ≤300 ppm
  • EP/BP - Ammonium (NH4):
    ≤0.02%
  • EP/BP - Iron (Fe):
    ≤10 ppm
  • EP/BP - Loss on Orying:
    ≤0.5%
  • EP/BP - Ash (sulfated):
    ≤0.1%
  • JP - Assay (C6H9N302) (dried basis):
    99.0 - 101.0%
  • JP - Ammonium (NH4):
    ≤0.02%
  • JP - Chloride (Cl):
    ≤0.021%
  • JP - Clarity and Color of Solution:
    Passes Test
  • JP - Iron (Fe):
    ≤10 ppm
  • JP - Identification:
    Passes Test
  • JP - Loss on Orying at 105 °C:
    ≤0.30%
  • JP- 0ptical Rotation [A)2°AO (+):
    11.8 - 12.8°
  • JP - Related Substances:
    Passes Test
  • JP - pH:
    7.0 - 8.5
  • JP - Residue on Ignition:
    ≤0.1%
  • JP - Sulfate (S04):
    ≤0.028%
  • ChP - Assay (dried basis):
    99.0 - 101.0%
  • ChP -Specific Rotation @ 20 °C (+):
    12.0 - 12.8°
  • ChP - Identification 1 (TLC):
    Passes Test
  • ChP - Identification 2 (IR):
    Passes Test
  • ChP - Acidity or alkalinity (pH):
    7.0 - 8.5
  • ChP - Transmittance of solution:
    ≥ 98.0%
  • ChP - Chlorides:
    ≤0.02%
  • ChP - Sulfates:
    ≤0.02%
  • ChP - Ammonium:
    ≤0.02%
  • ChP - 0ther amino acids (TLC)(<=0.5%, one spot only):
    Passes Test
  • ChP - Loss on drying @ 105 °C:
    ≤0.2%
  • ChP - Residue on ignition:
    ≤0.1%
  • ChP - Iron:
    ≤0.001%
  • ChP - Heavy metals:
    ≤0.001%
  • ChP - Arsenic:
    ≤0.0001%
  • ChP - Bacterial endotoxin, EU/g:
    ≤6.0