ContactUsLinkComponent
HyClone™ CDM4PERMAb PER.C6 Media, HyClone products (Cytiva)
HyClone™ CDM4PERMAb PER.C6 Media, HyClone products (Cytiva)
Catalog # 76462-540
Supplier:  Cytiva
CAS Number:  
undefined
HyClone™ CDM4PERMAb PER.C6 Media, HyClone products (Cytiva)
Catalog # 76462-540
Supplier:  Cytiva
Supplier Number:  SH30872.01
CAS Number:  
Specifications
  • Media Format:
    Powder
  • Description:
    CDM4PERMAb medium without L-glutamine, phenol red
  • Size:
    5 L
  • Cat. No.:
    76462-540
Specifications
About this item
HyClone™ CDM4PERMAb is a high-performance cell culture medium developed to increase process yields in the production of human antibodies and recombinant proteins using PER.C6 (Crucell NV) technology. Manufactured according to cGMP guidelines.
  • Animal-derived component free (ADCF)
  • Chemically-defined formulation
  • Metabolically designed for high cell yield and recombinant protein production
  • Allows for direct or sequential adaptation