4-(Dimethylamino)cinnamic acid 98%
About this item
: for Research Use Only
- CAS Number:1552-96-1
- Formula:C₁₁H₁₃NO₂
- MDL Number:MFCD00004397
- Molecular Weight:191.23 g/mol
- Boiling Pt:360.3 °C (760 mm Hg)
- Melting Pt:225…228 °C (760 mm Hg)
- Melting Pt:225…228 °C at N/A mmHg
- Flash Pt:171.7 °C
- Storage Temperature:Ambient
5 Options Available Below
- Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Return Policy
Product Details & Documents
: for Research Use Only
- CAS Number:1552-96-1
- Formula:C₁₁H₁₃NO₂
- MDL Number:MFCD00004397
- Molecular Weight:191.23 g/mol
- Boiling Pt:360.3 °C (760 mm Hg)
- Melting Pt:225…228 °C (760 mm Hg)
- Melting Pt:225…228 °C at N/A mmHg
- Flash Pt:171.7 °C
- Storage Temperature:Ambient
Product Family Options
Product Information
- Pack typeSizeAvailabilityPrice
- Glass bottle1 gDiscontinued-Supplier Number: -Item requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.Specifications:MarketsourceMarketsource
- Supplier Number: A254877-1GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Glass bottle5 gDiscontinued-Supplier Number: -Specifications:MarketsourceMarketsource
- Supplier Number: A254877-5GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A254877-10GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A254877-25GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
- Supplier Number: A254877-100GItem requires temperature control for storage and delivery with additional fees. It's not eligible for return due to safety and quality concerns. Consider requirements before purchasing.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



