Specifications
- Description:Keck sieve shaker kit, stainless steel screens, acrylic cylinders, ABS frame
- Cat. No.:MSPP-EW5997810
Specifications
About this item
Lightweight, compact design, easy to take into the field.
- Kit includes 20 interchangeable screens in a wide range of mesh sizes
- Provides clear indication of sample
- A complete kit with proven, reliable results
This complete mechanical sieve kit provides reliable particle size analysis. Kit includes 20 stainless steel mesh screens and five interlocking cylinders. Shakers are easy to use; place the sample in the top cylinder and shake to distribute particles, the volumetric percentages are indicated on the shaker frame. The clear acrylic cylinders make it easy to see sample and accurately read the volume. Each cylinder measures 1.85L×2" Int.Ø; ABS frame measures 14¹/₂L×4" Int.Ø.
Sieve szes included:
0.187" opening (No. 4 U.S. standard)
0.132" opening (No. 6 U.S. standard)
0.090" opening (No. 8 U.S. standard)
0.072" opening (No. 10 U.S. standard)
0.060" opening (No. 12 U.S. standard)
0.051" opening (No. 14 U.S. standard)
0.046" opening (No. 16 U.S. standard)
0.040" opening (No. 18 U.S. standard)
0.030" opening (No. 20 U.S. standard)
0.026" opening (No. 25 U.S. standard)
0.023" opening (No. 30 U.S. standard)
0.020" opening (No. 35 U.S. standard)
0.015" opening (No. 40 U.S. standard)
0.009" opening (No. 60 U.S. standard)
0.0055" opening (No. 100 U.S. standard)
0.0046" opening (No. 120 U.S. standard)
0.0041" opening (No. 140 U.S. standard)
0.0029" opening (No. 200 U.S. standard)
0.0024" opening (No. 230 U.S. standard)
0.0021" opening (No. 270 U.S. standard)