Order Entry
Puerto Rico
ContactUsLinkComponent
Quidel QuickVue Influenza A+B Test, NDC
Quidel QuickVue Influenza A+B Test, NDC
Catalog # 76474-226
CAS Number:  
undefined
Quidel QuickVue Influenza A+B Test, NDC
Catalog # 76474-226
Supplier Number:  QUI 20183
CAS Number:  

Specifications

  • Description:
    QuickVue influenza A+B test
  • Cat. No.:
    76474-226

Specifications

About this item

The QuickVue Influenza A+B Test detects and differentiates influenza type A and type B antigens directly from nasal swab and nasopharyngeal swab specimens.

  • PPA - A: 81.5%, B: 80.9%
  • NPA - A: 97.8%, B: 99.1%
  • CLIA complexity: waved

A single sample can be used to run both the QuickVue Influenza A+B Test and QuickVue RSV10 Test.

Processing time: 10 minutes or less