ContactUsLinkComponent
Oakton® EC200 Conductivity Cell for Oakton® 200 Series, Cole-Parmer
  76471-966
undefined
Oakton® EC200 Conductivity Cell for Oakton® 200 Series, Cole-Parmer
  76471-966
 :  35660-58
  • Ø:
    1.6 cm
  • Temperature range:
    0...80 °C (32...176 °F)
  • Description:
    Oakton® EC200 conductivity cell, K=1, BNC and phono
  • Length:
    15 cm
  • Environmentally Preferable:
  • Cat. No.:
    76471-966
  • Measurement range:
    0.1 mS/m – 10 S/m
  • For use with:
    Oakton 200 series meters and meter kits
  • Cable length:
    1 m
EC200 conductivity cell for Oakton® 200 series meters and meter kits.