Neogen® Colitag™ Presence/Absence (P/A) Water Test Kit, Hardy Diagnostics
Catalog # 76446-890
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Description:Presence/Absence (P/A) water test kit
- Cat. No.:76446-890
Specifications
About this item
The strength of the Colitag™ water test kit lies in its ease of use.
- Enhanced blue fluorescence to eliminate questionable positive E. coli results
- Detects one CFU of total coliform or E. coli bacteria per 100 ml water sample
- Resuscitates chlorine-injured bacteria
- Growth inhibitors to provide selectivity without affecting recovery of target organisms
Simply add Colitag™ to a 100 ml sample, incubate between 16 and 48 h, use a simple visual check for coliforms in the first instance, then a fluorescence check to determine E.coli bacteria presence.
Certifications: US EPA approved for MPN.