Orders LinkContactUsLinkComponent
TRIO.BAS™ Air Sampler Software PC
TRIO.BAS™ Air Sampler Software PC
Software PC
Catalog # 76446-766
Supplier:   Hardy Diagnostics
Supplier #:  BAS296
TRIO.BAS™ Air Sampler Software PC
Catalog # 76446-766
Supplier:  Hardy Diagnostics
TRIO.BAS™ Air Sampler Software PC
TRIO.BAS™ Air Sampler Software PC
Software PC
Price
$5,020.00
Log in for availability
Each
Minimum order quantity is 1
Product Details & Documents
BAS Software allows the transfer of environmental sampling data from a bluetooth or data transfer cable equipped TRIO.BAS™ instrument to and from a personal computer. The software allows database management, editing, reporting, and remote control of the instrument.
  • Management of users, passwords, and related permissions
  • Communication and data transfer between TRIO.BAS™ air samplers
  • Collected data cannot be modified according to CFR
  • ALCOA compliant
  • Permits input of other environmental sampling information e.g: surface, passive air sampling (settling plates), and compressed air/gas sampling
  • Upload sample photos when used with the CFU photo camera 76470-206
The software allows data base management, editing, reporting and remote control of the instrument.
Certifications: ALCOA compliant.
Ordering information: TRIO.BAS™ instrument must be configured with firmware 7.xx.
Specifications
  • Description:
    Software PC