Orders LinkContactUsLinkComponent
MAC X-ray Microanalysis Individual Standards, Singles, Electron Microscopy Sciences
Highly alloyed SINGLE
  76468-748
 :  80074-BAS-SS
MAC X-ray Microanalysis Individual Standards, Singles, Electron Microscopy Sciences
  76468-748
 :  
MAC X-ray Microanalysis Individual Standards, Singles, Electron Microscopy Sciences
MAC X-ray Microanalysis Individual Standards, Singles, Electron Microscopy Sciences
Highly alloyed SINGLE
 :  
$586.17
Each
All MAC X-ray Microanalysis standards are suitable for electron-beam energy dispersive or wavelength dispersive X-ray microanalysis systems.
All the standards are polished to a 0.25 µ diamond finish and carbon coated. Supplied in 2, 3, and 5 mm diameter and a location map for standard identification. They are supplied with fully authenticated certificates of analysis.
  • Description:
    Highly alloyed SINGLE