ContactUsLinkComponent
HardyDisks™ AST Sulfisoxazole, G-0.25, Hardy Diagnostics
HardyDisks™ AST Sulfisoxazole, G-0.25, Hardy Diagnostics
Catalog # 76443-246
Supplier:  Hardy Diagnostics
CAS Number:  
undefined
HardyDisks™ AST Sulfisoxazole, G-0.25, Hardy Diagnostics
Catalog # 76443-246
Supplier:  Hardy Diagnostics
Supplier Number:  Z9075
CAS Number:  
Specifications
  • Pack type:
    1×50 disks
  • Description:
    Test disks
  • Cat. No.:
    76443-246
Specifications
About this item
HardyDisk™ AST’s are impregnated paper disks used for Antimicrobial Susceptibility Testing (AST); also known as disk diffusion or Kirby-Bauer testing.
Each cartridge contains 50 disks that are spring loaded for smooth and reliable dispensing.
Delivery: Freeze on arrival.