Order Entry
Puerto Rico
ContactUsLinkComponent
HyClone Cell Boost™ 7a Supplement, HyClone products (Cytiva)
HyClone Cell Boost™ 7a Supplement, HyClone products (Cytiva)
Supplier:  Cytiva
Certificates
undefined
HyClone Cell Boost™ 7a Supplement, HyClone products (Cytiva)
Supplier:  Cytiva

Specifications

  • Storage Conditions:
    Away from light and moisture
  • Shelf Life:
    18 months
  • Storage Temperature:
    2…8 °C
  • Composition:
    Without: L-glutamine
    With: Poloxamer 188

Specifications

Related Information