Fluid D with Tween, Hardy Diagnostics
Catalog # 76443-236
Supplier: Hardy Diagnostics
CAS Number:
Specifications
- Pack type:PP bottle, 125 ml
- Description:Fluid D with Tween
- Size:12/pack
- Cat. No.:76443-236
- Fill volume:100 ml
- Volume:125 ml
Specifications
About this item
Fluid D is used by the pharmaceutical and medical device industry in procedures used to ensure sterility of injectables, non-injectables, liquid products, oils and ointments, antibiotic powders, sterile pathway devices and aerosols.
- Bottles are tough and sturdy to reduce breakage
- Secure tight lid ensures product integrity
- Optically clear bottles are made of clarified polypropylene for clear viewing
- Bottles have excellent impact and chemical resistance