Order Entry
Puerto Rico
ContactUsLinkComponent
AccuWipe® Recycled One-Ply Delicate Task Wipers, Georgia Pacific®
AccuWipe® Recycled One-Ply Delicate Task Wipers, Georgia Pacific®
Catalog # 76469-042
undefined
AccuWipe® Recycled One-Ply Delicate Task Wipers, Georgia Pacific®
Catalog # 76469-042
Supplier Number:  GPC29712

Specifications

  • Color:
    White
  • Material:
    Tissue
  • Cat. No.:
    76469-042
  • Packaging:
    280/box
  • W×L:
    4.5×8.25"

Specifications

About this item

Soft, non-abrasive, low-lint and absorbent wipers can be used when a gentle touch is needed.

  • Strong and even more gentle when used wet
  • Anti-static poly-shield to minimize electrostatic discharge

Choose for delicate or critical cleaning jobs of lenses, precision parts, and instruments.