Pacific Blue Select™ Disposable Patient Care Washcloths, Georgia Pacific®
Catalog # 76469-036
Supplier: Janitorial Supplies
Specifications
- Color:White
- Material:Cellulose, bonded
- Cat. No.:76469-036
- Packaging:55/pack, 24 packs/carton
- W×L:10×13"
Specifications
About this item
Pacific Blue Select™ A400 disposable patient care washcloths are premium-weight personal disposable washcloths.
- Soft airlaid bonded cellulose has a bulky texture when wet, so it's soft on skin and easy to open when rinsed and wrung out
- Durable construction stands up in light solvents
- Convenient dispenser packs help protect towels from moisture and contamination
- Economical choice for patient care, healthcare, and light manufacturing cleaning tasks
- Absorbent material soaks up spills quickly
Specifically designed with softness in mind for personal care use, convenient single-ply Pacific Blue Select™ disposable washcloths for adults are thick and absorbent. They stay extremely soft and bulky when wet for extended use.
Made from airlaid bonded cellulose, these disposable bathroom towels deliver an excellent premium cloth replacement wiper for patient care and patient room cleaning.