Chix® Pro-Quat Fresh Guy® Food Service Towels, Heavy-Duty, Chicopee®
Catalog # 76469-020
Supplier: Janitorial Supplies
Specifications
- Color:Red
- Material:Paper
- Environmentally Preferable:
- Cat. No.:76469-020
- Packaging:150 wipes/pack
- W×L:12.5×17"
Specifications
About this item
Keeping a professional kitchen clean to support food safety initiatives can be an arduous task, but these food service towels will surely make the job easier.
- Absorbent, nonwoven fabric is low-linting and durable
- Can be machine washed or rinsed and reused
Engineered to release full strength Quat sanitizer on the surface, reducing the risk of cross-contamination that can lead to food borne illness outbreaks. Made with Microban® antimicrobial protection, which inhibits the growth of odor-causing bacteria and mold on the product. Best of all, because the antimicrobial formula is built in to the product, it won't simply wash off. Heavy-duty design with Fresh Guy® pattern features larger apertures for increased particle pickup.